Learn More
StatLab MasterTech PTAH Stain Kit - Microwave
Catalog No. 23640009
Change view
Click to view available options
Quantity:
100 mL
473 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23640009 100 mL
23640010 473 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23640009 Supplier StatLab Supplier No. KTPTA
Only null left
Harmful if swallowed, in contact with skin, or if inhaled
Stains for collagen, fibrin, and striated muscle in under one hour
  • This kit includes a 55 minute microwave procedure and the traditional 15 hour procedure.
  • Unlike similar kits on the market, the MasterTech P.T.A.H. Stain Kit is less hazardous, using Zinc Chloride instead of mercuric based mordants, which also eliminates the need for Potassium Dichromate.
  • Ferric Ammonium Sulfate is also used instead of Acidic Potassium Permanganate, further reducing the stain kits hazard. P.T.A.H. serves as the primary stain, producing blue fibrin, muscle, and nuclei, with brown to red collagen.
  • A MasterTech Special Stain Kit.
Specifications
Product Type Special Stain Kit
Quantity 100 mL