missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech Periodic Acid Reagent

Catalog No. 21402155
Change view
Click to view available options
Quantity:
25 gm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21402155 25 gm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21402155 Supplier StatLab Supplier No. SPP0377
Only null left
Harmful if swallowed, in contact with skin, or if inhaled

Acid reagent, used to detect carbohydrates and carbohydrate-rich compounds in tissues

  • Formulation is particularly useful for identifying glycogen, mucin, glycoproteins, and basement membranes
  • The reagent works by oxidizing carbohydrates, exposing aldehyde groups, and then reacting with the Schiff reagent to produce a magenta color, which indicates the presence of carbohydrates in the tissue
  • This staining technique is invaluable in diagnosing various diseases, including fungal infections, glycogen storage disorders, and certain types of cancers

Specifications

Physical Form Crystalline Powder
Color White
CAS 10450-60-9
Quantity 25 gm
Grade Laboratory