missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MasterTech Oxidizer Solution, StatLab
Description
- This solution is added to hematoxylin to oxidize it into hematein the active ingredient in the staining solution
- The oxidation process is crucial for the staining of tissue samples as it allows the dye to bind to the negatively charged phosphate groups of nuclear chromatin resulting in the characteristic blue-purple color of the stain
- The use of sodium iodate as an oxidizer is common and widely accepted in histology laboratories
- A StatLab MasterTech Special Stain Kit Component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 64-18-6, 108-95-2, 7647-01-0, 64-17-5 |
| Color | Colorless |
| Quantity | 473 mL |
| Grade | Laboratory |