missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab MasterTech Oil Red O Stain
Description
- Stain is a fat-soluble diazo dye that functions as a pigment and is classified as one of the Sudan dyes
- It is particularly useful in diagnostic, research, and forensic purposes, as it helps in identifying conditions where fat may appear in abnormal locations, such as fat emboli or tumors arising from fat cells
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| CAS | 57-55-6 |
| Grade | Laboratory |
| Color | Red |
| Quantity | 16 oz. |