Learn More
StatLab MasterTech H and E Stain Kit
Description
- For more than three decades, the MasterTech brand has been known for its premium Harris Hematoxylin
- Only dyes certified by the Biological Staining Commission are used in MasterTech Harris Hematoxylin ensuring that every batch of Hematoxylin provides consistent and vibrant nuclear staining
- Harris Hematoxylin serves as the primary stain, producing blue to purple nuclei against a pink to red background, counterstained by Eosin Y
- Pint Stain Kit yields between 320-500 slides
- Pint Kit Consists of: Harris Hematoxylin pint, Eosin Y Stain pint, Differentiating Solution pint, and Bluing Solution pint
- A MasterTech Special Stain Kit
Specifications
Specifications
| Type | Hematoxylin and Eosin |
| Product Type | Special Stain Kit |