missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab MasterTech 5% Chromic Acid
Description
- A component for the MasterTech GMS Stain Kit
- This staining method provides a detailed view of the tissue, allowing pathologists to identify fungal infections and other abnormalities
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Quantity | 1000 mL |
| Physical Form | Solution |