missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech 3% Ferric Ammonium Sulfate

Catalog No. 21033384
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21033384 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21033384 Supplier StatLab Supplier No. KC3931

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a mordant specifically in the staining process

  • Used as a mordant, specifically in the staining process, where it helps to bind the dye to the tissue by increasing its affinity to certain cellular components
  • A component for the MasterTech Chandlers Precision Reticulum Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Ferric Ammonium Sulfate
CAS 7783-83-7
Physical Form Liquid or Solid
Quantity 100 mL
Percent Purity 3%
Grade Laboratory
Packaging Plastic bottle