Learn More
Masterflex™ L/S™ High-Performance Precision Pump Tubing, Platinum-Cured Silicone
Excellent biocompatibility that meets multiple classifications
$897.47 - $1,016.88
Specifications
Autoclavable Autoclavable
Material Platinum-cured Silicone
View More Specs
Products 2
Catalog Number Mfr. No. Tubing Size Price Quantity & Availability  
Catalog Number Mfr. No. Tubing Size Price Quantity & Availability  
13200287
View Documents Promotion Details
Masterflex™
MFLX9641035
35
Each for $897.47
Only null left
13200288
View Documents Promotion Details
Masterflex™
MFLX9641036
36
Each for $1,016.88
Only null left
Description
Description
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Peroxide-Cured Silicone High-Performance Precision Pump Tubing provides accurate, repeatable flow. Ideal for pharmaceutical and biotechnology, peristaltic pumping and food applications.
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock — most formulations and sizes available to ship same day
  • Economical tubing with a longer tubing life
Specifications
Specifications
Autoclavable
Platinum-cured Silicone