Learn More
Masterflex™ L/S™ Compact Variable-Speed Drives
Masterflex™ L/S™ Compact Variable-Speed Drives makes low-flow pumping easy.
$3,762.72 - $3,836.02
Specifications
Control Type Analog
Display Type No Display
Reversible Yes
Includes L/S Miniflex pump head and dual-voltage (115/230 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.
Material Painted Steel
View More Specs
Products 2
Catalog Number Mfr. No. Flow Rate Channels Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Channels Price Quantity & Availability  
13200003
View Documents Promotion Details
Masterflex™
MFLX7724020
1.6 to 239 mL/min per channel 1
Each for $3,762.72
Only null left
13200004
View Documents Promotion Details
Masterflex™
MFLX7724030
1.6 to 124 mL/min per channel 2
Each for $3,836.02
Only null left
Description
Description
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
  • Separate single-turn speed control and forward/off/reverse switch—maintain speed setting when turning pump on or off
  • Remote start/stop via contact closure on back of pump
Note:  Tubing is not included; sold separately.
Specifications
Specifications
Analog
Yes
Painted Steel
90/260 VAC
No Display
L/S Miniflex pump head and dual-voltage (115/230 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.
90 to 260 VAC, 50/60 Hz