Learn More
Master Lock™ Safety Lockout Padlocks
Lightweight Zenex thermoplastic body
Supplier:  Master Lock™ 410PRP
Encompass
Catalog No. 189991350G
Description
Description
  • 6-pin reserved for safety cylinder with over 40,000 key changes
  • Non-rekeyable, key retaining
  • Write-on English, Spanish and French labels included
  • 1-3/4 in. body height
  • 1-1/2 in. body width
  • Shackle (Dia. × W × H): 1/4 in. × 25/32 in. × 1-1/2 in.
Specifications
Specifications
Purple
0.635 cm
1.5 in.
3.8 cm
6
0.78 in.
1.98 cm
0.25 in.
1.75 in.
4.45 cm
Write-on English, Spanish, and French labels
1.5 in.
3.8 cm
Product Suggestions
Product Suggestions