Learn More
Master Lock™ Safety High-Visibility Aluminum Padlock
Supplier:  Master Lock™ 411GRN
Encompass
 View more versions of this product
Catalog No. 296353400
Description
Description
Withstands harsh, corrosive environments; available in 6 vibrant colors; Over 5-pin cylinder with over 10,000 key changes
  • Durable padlock protection with a high-visibility, powder-coated finish
  • Vibrant colors provide safety color recognition plus high security
  • These aluminum locks have a pick-resistant five-pin Pro Series™ cylinder, but are conveniently rekeyable Corrosive-resistant locks are all 1 1/2 in. wide (40mm) and have shackle diameters of 1/4 in. (6.35mm)
  • Vertical shackle clearance is 1 1/16 in. (27mm)
  • Uses FDA approved lubricants safe for use in food processing
Product Suggestions
Product Suggestions