Learn More
Brady™ Marking Tape Roll: High Performance Laminated Vinyl, Diagonal Stripes, Red on White
Catalog No. 19103759
Change view
Click to view available options
Color:
Red on White
Catalog No. Color
19103759 Red on White
1 options
Catalog No. 19103759 Supplier Brady™ Supplier No. 55295
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High Performance Laminated Vinyl (B-950) tape is perfect for marking obstructions, low hanging objects, pillars, passageways, aisles and other hazardous areas or objects.
  • High gloss, durable vinyl with pressure-sensitive rubber-based adhesive withstands most common chemicals, solvents, oils and water
  • Encapsulated graphics are sealed beneath a protective layer of abrasion-resistant vinyl
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Designed to withstand tough indoor conditions
Specifications
Adhesive Type Rubber-based Pressure-Sensitive
Color Red on White