missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Marbles

Catalog No. S96595
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96595 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96595 Supplier Eisco™ Supplier No. PMBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Plastic Marbles for educational puzzles and games.

  • 5 marbles per pack.

Specifications

Grade K to 12
Format Kit
Product Type Marbles
Quantity 5 Pk.
Material Plastic
For Use With (Application) Classroom Activities and Demonstration
Subject Physics - Motion
Length (English) 2 in.
Length (Metric) 5 cm