missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Steel Marbles

Catalog No. S96594
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96594 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96594 Supplier Eisco™ Supplier No. SMBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Steel Marbles for educational puzzles and games.

  • 5 marbles per pack.

Specifications

Grade Level High School
Grade 6 to Undergraduate
Height (English) 1 in.
For Use With (Application) Classroom activities
Includes 5 Marbles
Material Steel
Quantity 5 Pk.
Weight (English) 0.4 lb.
Weight (Metric) 181.43 g
Length (English) 10 in.
Class Size 5 Lab Groups
Product Type Curriculum Set
Show More Show Less