missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPA™ ChemZoil™ NL339 Chemical Resistant Neoprene Gloves
Neoprene multi-layer gloves provide chemical resistance against acids and alkalis, such as concentrated hydrofluoric acid, oils, and greases.
$109.08 - $109.08
Specifications
| Certifications/Compliance | 21 CFR 170-199 |
|---|---|
| Certifications/Compliance (Europe) | Cat III CE 0334, EN 407:X1XXXX, EN 511 |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-046-580
|
MAPA™
339429 |
9 |
Case of 6 Pair for $109.08
|
|
|||||
|
19-046-582
|
MAPA™
339421 |
11 |
Case of 6 Pair for $109.08
|
|
|||||
Description
- Protects against acids
- For use with chemical protective suits
- Black, heavyweight, neoprene
- Articulated finger design for added comfort, flexibility and reduced hand fatigue
- Soft, interlocked knit cotton liner absorbs perspiration and adds comfort
- Multilayer construction assures a leakproof glove
- Roughened grip with pinked cuff
- Length: 14 in. (35.6 cm)
Specifications
| 21 CFR 170-199 | |
| Black | |
| Neoprene | |
| 1.20 mm | |
| Yes | |
| Bacteria, Fungi only | |
| Pinked | |
| Cotton Knit | |
| ChemZoil™ NL-339 | |
| 35.5 cm |
| Cat III CE 0334, EN 407:X1XXXX, EN 511 | |
| Yes | |
| 50 mil | |
| EN 388:3121 | |
| ABCJKL | |
| Chemical processing, alkaline units at petroleum refineries, metal plating operations, hazardous material handling, hazmat and chemical spills | |
| Pebbled | |
| Lined | |
| 14 in. |