missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ TempDex 710 Sandy Nitrile-Coated Thermal Protection Gloves

Catalog No. 19046552
Change view
Click to view available options
Size:
11
7
9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19046552 11
19046550 7
19046551 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19046552 Supplier MAPA™ Supplier No. 34710031

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thin, anatomically-shaped gloves ensure dexterity and provide thermal protection from temperatures up to 150°C.

Thermal protection is now available for precision work

  • Silicone free to prevent traces or defects on metal sheet and glass prior to painting
  • Thin, anatomically-shaped design ensures dexterity
  • Dot embossing improves grip and insulation
  • Suitable for long-term wear

Specifications

Coating Material Sandy Nitrile
Color Yellow
Size 11
Temperature (English) Max. 300°F
Temperature (Metric) Max 150°C
For Use With (Application) Construction, Ceramics, Heat Processing, and Pipe/Steel Handling
Glove Cuff Style Knit
Series TempDex 710