Learn More
MAPA™ KryTech 395 Heavy Duty Work Gloves
Description
- Combined protection against cuts, liquid, and contact heat up to 100°C
- Nitrile coating enhances grip capabilities in wet or slippery applications
- Entirely liquid-proof, withstanding mechanical aggression
- Micro-organism protection against bacteria and fungi
- Silicone-free to prevent traces and defects on metal sheets or glass
- Proven long-lasting performance
Specifications
Specifications
| Color | Green |
| Cut Resistance Rating | Level D |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Cut-resistant Gloves |
| Size | 9 |