Learn More
MAPA™ Krytech 579 Precision Work Gloves
Low risk, ISO B gloves provide standard protection for intricate handling in reasonably clean environments.
Supplier:  MAPA™ 34579088
 View more versions of this product
Catalog No. 19-249-763
Only null left
Explore available promotions
Description
Description
  • Ergonomic shape for the reduction of hand fatigue
  • Abrasion resistance for long-term wear
  • Reduced thickness for tactile sensitivity
  • Anatomical shape for dexterity
  • High density polyurethane fiber interior
  • Ventilated back
  • White
Applications:
Automotive/mechanical industry; Paper industry
Specifications
Specifications
Cat. 2 CE, EN 420
ANSI A4
Cut-resistant Gloves
EN 388:4342B
Knit
KryTech 579
24.1 cm
White, Gray
HDPE
8
Handling and sorting small sharp parts, finishing work
579128
9.5 in.
Product Suggestions
Product Suggestions