missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ TriLites 984 Extended Length Tripolymer Gloves

Catalog No. 19165718
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165718 X-Large
19165715 Small
19165716 Medium
19165717 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19165718 Supplier MAPA™ Supplier No. 34984029
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Extra long, 11.5 in. length offers extended protection for the wrist and upper forearm when working with chemicals.

Tripolymer blend of neoprene, nitrile and natural rubber provides comfort of latex exam gloves with added strength, tear resistance and chemical splash protection

  • Exclusive finish minimizes tape and label sticking to gloves
  • Textured grip with lightly halogenated finish for optimum tactility and grip; wet or dry
  • Ambidextrous design, rolled cuff, unlined
  • Powder-free, clean process (CP) for no cross contamination
  • Thickness: 6 mil
  • Length: 11.5 in. (29 cm)
  • Purple

Applications:

Laboratory research, light assembly and sorting, precision parts handling, light maintenance

Specifications

Certifications/Compliance ASTM D3578, 21 CFR 170-199
Certifications/Compliance (Europe) Cat 3 CE 0334
Chlorinated Yes
Color Purple
Hand Ambidextrous
Latex Free No
Material Tripolymer
Powdered Powder Free
Size X-Large
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Laboratory research, light assembly and sorting, precision parts handling, light maintenance
Glove Cuff Style Rolled
Grip Design Pebbled
Lining Unlined
Model 984929
Series TriLItes 984
Length (English) 11.5 in.
Length (Metric) 29.2 cm
Show More Show Less