missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP3K10 (MLK2), GST Tag Recombinant Protein

Catalog No. PV3877
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3877 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3877 Supplier Invitrogen™ Supplier No. PV3877
Only null left

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

MAP3K10 (MLK2) is a serine/threonine kinase which has been shown to activate MAPK8/JNK and MKK4/SEK1. This kinase itself can be phoshorylated, and is thus activated by JNK kinases. MAP3K10 functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.

Specifications

Accession Number Q02779
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 2mM DTT, 150mM NaCl, 0.02% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 4294
Molecular Weight (g/mol) 131.2 kDa
Name Human MAP3K10 (MLK2), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BC028668; BC046514; Map3k10; MAP3K9; MEKK10; MEKK9; mitogen activated protein kinase kinase kinase 10; mitogen-activated protein kinase kinase kinase 10; Mixed lineage kinase 2; mixed-lineage kinase 2; MKN28 derived nonreceptor_type; MKN28 derived nonreceptor_type serine/threonine kinase; MKN28 kinase; MLK1; MLK2; MST; PRKE1; protein kinase MST; serine/threonine kinase
Common Name MLK2
Gene Symbol MAP3K10
KinaseFamily MLK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Assay Entry Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Omnia™ Bulk Substrate
Target Entry MAP3K10
Target Gene mitogen-activated protein kinase kinase kinase 10
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant
Form Liquid
Protein Subtype Serine⁄Threonine Kinases
Research Category Signal Transduction
Purity or Quality Grade 60% by SDS-PAGE
Show More Show Less