missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slice™ Manual Small Scraper
The Slice™ 10594 Manual Small Scraper uses a thinner, narrower blade than our larger utility scrapers.
Supplier: Slice™ 10594
Description
- Installed blade type: Rounded-tip No. 10524
- Durable nylon handle with textured slider
- Manual retraction with 1 scraping position
- Ambidextrous design
- Finger-friendly™ blade scrapes effectively
- Reduces injuries, lowers costs
- Non-sparking, non-conductive, non-magnetic blade
- Chemically inert blade never rusts
- Oil- and lubricant-free blade
- 100 percent zirconium oxide blade
- Blade edge lasts up to 11.2x longer than metal
- Fewer blade changes = fewer injuries
- No-tool blade change
Specifications
| Scraper | |
| Compatible blades: 10524, 10525, 10523 | |
| 63.2 x 67.0 x 10.7 mm |
| Zirconium Oxide | |
| GFN, POM, Carbon Steel | |
| 18.0 g |