missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannose triflate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 118.0°C to 122.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (ELSD) (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Solubility Information | Clear colorless to slightly yellow |
| Formula Weight | 480.36 |
| Percent Purity | 98% |
| Chemical Name or Material | Mannose triflate |
RUO – Research Use Only