missing translation for 'onlineSavingsMsg'
Learn More

Manganese telluride, 99.9% (metals basis)

Catalog No. AA3638909
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3638909 10 g
AA3638903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3638909 Supplier Thermo Scientific Chemicals Supplier No. 03638909
Only null left

CAS: 12032-88-1 | MnTe | 182.54 g/mol

Manganese telluride is used in scientific research and developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12032-88-1
Molecular Formula MnTe
Molecular Weight (g/mol) 182.54
MDL Number MFCD00049487
InChI Key VMINMXIEZOMBRH-UHFFFAOYSA-N
Synonym manganese telluride, telluroxomanganese, manganese monotelluride, manganous telluride, manganese ii telluride, manganese 2+ telluride
PubChem CID 82828
IUPAC Name tellanylidenemanganese
SMILES [Mn]=[Te]

Specifications

Color Gray
Physical Form Lump
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 182.54
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Manganese telluride
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 234-782-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only