missing translation for 'onlineSavingsMsg'
Learn More

Manganese powder, -140+325 mesh, 99.6% (metals basis)

Catalog No. p-7051494
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4134314 25 g
AA4134322 100 g
AA4134336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4134314 Supplier Thermo Scientific Chemicals Supplier No. 04134314
Only null left

CAS: 7439-96-5 | Mn | 54.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Quantity 25 g
Form Powder
UN Number UN3089
Percent Purity 99.6%
Assay Metals basis
Chemical Name or Material Manganese powder
Hazard Category H228-H319
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-105-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only