missing translation for 'onlineSavingsMsg'
Learn More

Manganese pieces, irregular, 99.95% (metals basis)

Catalog No. p-4781988
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3622114 25 g
AA3622122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3622114 Supplier Thermo Scientific Chemicals Supplier No. 03622114
Only null left

For Vacuum Deposition | CAS: 7439-96-5 | Mn | 54.94 g/mol

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Color Gray
Appearance Gray irregularly shaped pieces
Density 7.3g/cm3 at 20°C
Quantity 25 g
Form Pieces
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Manganese
EINECSNumber 231-105-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only