Learn More
Manganese nanopowder, APS 30-50nm
Form: Mn3O4 layer 5-12nm | CAS: 7439-96-5 | Mn | 54.94 g/mol
Supplier: Thermo Scientific Chemicals 04550814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7439-96-5 | |
| 25 g | |
| Nanopowder | |
| MFCD00011111 | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| manganese | |
| 23930 |
| Manganese | |
| Gray powder | |
| 99.9% (Metals basis) | |
| Mn | |
| colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
| [Mn] | |
| 54.94 | |
| CHEBI:35154 |
Chemical Identifiers
| 7439-96-5 | |
| 54.94 | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| 23930 | |
| manganese |
| Mn | |
| MFCD00011111 | |
| colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
| CHEBI:35154 | |
| [Mn] |
Safety and Handling
Take precautionary measures against static discharges.
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-105-1
TSCA : Yes
RUO – Research Use Only