missing translation for 'onlineSavingsMsg'
Learn More

Manganese nanopowder, APS 30-50nm

Catalog No. AA4550806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4550806 5 g
AA4550814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4550806 Supplier Thermo Scientific Chemicals Supplier No. 04550806

Form: Mn3O4 layer 5-12nm | CAS: 7439-96-5 | Mn | 54.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Color Gray
Appearance Gray powder
Density 7.3g/cm3 at 20°C
Quantity 5 g
Form Nanopowder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Manganese
Hazard Category H228-H319
Hazard Statement Take precautionary measures against static discharges.
Precautionary Statement P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-105-1
TSCA Yes

RUO – Research Use Only