missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

Supplier:  Thermo Scientific Chemicals 01080522

 View more versions of this product

Catalog No. AA1080522


Only null left
Explore available promotions

Description

Description

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(IV) oxide
1313-13-9
Powder
99.997% (Metals basis)
MFCD00003463
Insoluble in water
O=[Mn]=O
86.94
86.94
5.026
535°C (decomposition)
Brown
100 g
MnO2
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
Odorless

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c

H302+H332-H373

EINECSNumber : 215-202-6

RTECSNumber : OP0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only