Learn More
Manganese(IV) oxide activated, tech. 90%
CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Supplier: Thermo Scientific Chemicals L1325322
Description
Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)Â acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsManganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)Â acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| Manganese(IV) oxide activated | |
| 1313-13-9 | |
| MnO2 | |
| 14,5730 | |
| Insoluble in water. | |
| O=[Mn]=O | |
| 86.94 | |
| 86.94 | |
| Technical |
| ∽535°C (decomposition) | |
| 100 g | |
| MFCD00003463 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| dioxomanganese | |
| 14801 | |
| 90% | |
| 5.026 |
Chemical Identifiers
| 1313-13-9 | |
| 86.94 | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| 14801 | |
| O=[Mn]=O |
| MnO2 | |
| MFCD00003463 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| dioxomanganese |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H373
EINECSNumber : 215-202-6
RTECSNumber : OP0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only