missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide activated, Honeywell Fluka™

Catalog No. p-7072247
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002799 250 g
6002798 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002799 Supplier Honeywell Chemicals Supplier No. 63548250G
Only null left

Technical, activated,≥90% (RT)

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Quantity 250 g
UN Number NONH for all modes of transport
Formula Weight 86.94g/mol
Percent Purity ≥90%
Chemical Name or Material Manganese(IV) oxide activated
RTECSNumber OP0350000