Learn More
Manganese(IV) oxide, 99.99+%, (trace metal basis)
CAS: 1313-13-9 | MnO2 | 86.94 g/mol
$147.16 - $495.94
Chemical Identifiers
| CAS | 1313-13-9 |
|---|---|
| Molecular Formula | MnO2 |
| Molecular Weight (g/mol) | 86.94 |
| MDL Number | MFCD00003463 |
| InChI Key | NUJOXMJBOLGQSY-UHFFFAOYSA-N |
| Synonym | manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein |
| PubChem CID | 14801 |
| IUPAC Name | dioxomanganese |
| SMILES | O=[Mn]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC193470050
|
Thermo Scientific Chemicals
193470050 |
5 g | Glass bottle |
Each for $147.16
|
|
||||
|
AC193470250
|
Thermo Scientific Chemicals
193470250 |
25 g | Glass bottle |
Each for $495.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagentChemical Identifiers
| 1313-13-9 | |
| 86.94 | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| 14801 | |
| O=[Mn]=O |
| MnO2 | |
| MFCD00003463 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| dioxomanganese |
Specifications
| 535.0°C | |
| Black to Gray | |
| 5 g | |
| MnO2 | |
| MFCD00003463 | |
| 15,5795 | |
| Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid | |
| O=[Mn]=O | |
| 86.94 | |
| 86.94 | |
| Trace Metal Basis | |
| 5.02g/mL |
| 1313-13-9 | |
| Powder, Crystals and/or Chunks | |
| 99.99+% | |
| MnO2 | |
| 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| dioxomanganese | |
| 14801 | |
| ≥99.99% | |
| Glass bottle | |
| Manganese(IV) oxide |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D
GHS Signal Word: Warning
EINECSNumber : 215-202-6
RUO – Research Use Only