Learn More
Manganese(IV) Oxide, 99%, -10 mesh
CAS: 1313-13-9 | MnO2 | 86.94 g/mol
$107.96 - $107.96
Chemical Identifiers
| CAS | 1313-13-9 |
|---|---|
| Molecular Formula | MnO2 |
| Molecular Weight (g/mol) | 86.94 |
| MDL Number | MFCD00003463 |
| InChI Key | NUJOXMJBOLGQSY-UHFFFAOYSA-N |
| Synonym | manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein |
| PubChem CID | 14801 |
| IUPAC Name | dioxomanganese |
| SMILES | O=[Mn]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC222581000
|
Thermo Scientific Chemicals
222581000 |
100 g | Plastic bottle |
Each for $107.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 1313-13-9 | |
| 86.94 | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| 14801 | |
| O=[Mn]=O |
| MnO2 | |
| MFCD00003463 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| dioxomanganese |
Specifications
| 535.0°C | |
| 100 g | |
| MnO2 | |
| MFCD00003463 | |
| 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 | |
| manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
| NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
| dioxomanganese | |
| 14801 | |
| 99% | |
| 5.02 |
| 1313-13-9 | |
| 99% | |
| MnO2 | |
| 1479 | |
| 15,5795 | |
| Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid | |
| O=[Mn]=O | |
| 86.94 | |
| 86.94 | |
| Plastic bottle | |
| Manganese(IV) oxide, 99+%, -100 mesh |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D
GHS Signal Word: Warning
EINECSNumber : 215-202-6
RTECSNumber : OP0350000
TSCA : TSCA
RUO – Research Use Only