missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(III) phosphate hydrate, 99%
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
It is employed as intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Specifications
Specifications
| Quantity | 10 g |
| Solubility Information | Insoluble in water. |
| IUPAC Name | manganese(3+);phosphate;hydrate |
| Formula Weight | 149.91 (Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | Manganese(III) phosphate hydrate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P301+P310-P305+P351+P338-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only