missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) fluoride, 98% (metals basis)

CAS: 7783-53-1

$98.49 - $386.60

Chemical Identifiers

CAS 7783-53-1
MDL Number MFCD00016223
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1306909
View Documents
Thermo Scientific Chemicals
01306909
10 g
Each for $98.49
Only null left
 
AA1306918
View Documents
Thermo Scientific Chemicals
01306918
50 g
Each for $386.60
Only null left
 
Description

Description

It is used as a fluorinating agent, useful for conversion of hydrocarbons into fluorocarbons. It is used in organic synthesis as Lewis acid to prepare a number of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-53-1
MFCD00016223

Specifications

(decomposition)
Crystalline
MFCD00016223
14,5741
111.93
Hygroscopic
Manganese(III) fluoride
7783-53-1
10 g
UN3288
Insoluble in organic solvents. Hydrolyzed by water.
98%
3.54 g/mL
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H272-H302+H332-H315-H318-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 232-006-6

RTECSNumber : OP0882600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only