Learn More
Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)
CAS: 15244-36-7 | H2MnO5S | 169.01 g/mol
$305.40 - $305.40
Chemical Identifiers
| CAS | 15244-36-7 |
|---|---|
| Molecular Formula | H2MnO5S |
| Molecular Weight (g/mol) | 169.01 |
| MDL Number | MFCD00149159 |
| InChI Key | ISPYRSDWRDQNSW-UHFFFAOYSA-L |
| Synonym | manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate |
| PubChem CID | 177577 |
| ChEBI | CHEBI:86364 |
| SMILES | O.[Mn++].[O-]S([O-])(=O)=O |
Description
Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15244-36-7 | |
| 169.01 | |
| ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
| 177577 | |
| O.[Mn++].[O-]S([O-])(=O)=O |
| H2MnO5S | |
| MFCD00149159 | |
| manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
| CHEBI:86364 |
Specifications
| 15244-36-7 | |
| Crystalline Powder | |
| 99.999% (Metal basis) | |
| MFCD00149159 | |
| Soluble in water. | |
| O.[Mn++].[O-]S([O-])(=O)=O | |
| 169.01 | |
| CHEBI:86364 | |
| Odorless | |
| Pink Crystalline Powder |
| Pink | |
| 25 g | |
| H2MnO5S | |
| manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
| ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
| manganese(2+) hydrate sulfate | |
| 177577 | |
| 151.00 (Anhydrous) | |
| Hygroscopic | |
| Manganese(II) sulfate hydrate, Loses all water at 400-450° |
Safety and Handling
P260-P314-P501c
H373
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 232-089-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only