missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) phthalocyanine

Catalog No. AA3930903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3930903 1 g
AA3930906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3930903 Supplier Thermo Scientific Chemicals Supplier No. 03930903

CAS: 14325-24-7 | C32H16MnN8 | 567.474 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14325-24-7
Molecular Formula C32H16MnN8
Molecular Weight (g/mol) 567.474
MDL Number MFCD00049821
InChI Key ICIFYHOILPYQKB-UHFFFAOYSA-N
PubChem CID 6508743
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2[N-]3.[Mn+2]

Specifications

Quantity 1 g
Solubility Information Soluble in pyridine,chloronaphthalene,and conc. sulfuric acid. Insoluble in water.
Formula Weight 567.47
Physical Form Crystalline
Chemical Name or Material Manganese(II) phthalocyanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only