missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxide, 99.99%, (trace metal basis)

Catalog No. p-4258693
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436970010 1 g Glass bottle
AC436970100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436970010 Supplier Thermo Scientific Chemicals Supplier No. 436970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1344-43-0 | MnO | 70.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 1344-43-0
Molecular Formula MnO
Molecular Weight (g/mol) 70.94
MDL Number MFCD00016226
InChI Key VASIZKWUTCETSD-UHFFFAOYSA-N
Synonym manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide
PubChem CID 14940
IUPAC Name oxomanganese
SMILES O=[Mn]

Specifications

Melting Point 1650.0°C
Color Green
Physical Form Crystalline Powder and/or Chunks
Quantity 1 g
Assay Percent Range 76.7 to 78.2% (Mn) (Complexometry)
Solubility Information Solubility in water: insoluble
Formula Weight 70.94
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Manganese(II) oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
EINECSNumber 215-695-8

RUO – Research Use Only