missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate tetrahydrate, 98%

CAS: 20694-39-7 | H8MnN2O10 | 251.006 g/mol

Supplier:  Thermo Scientific Chemicals A185210B

 View more versions of this product

Catalog No. AAA185210B


Only null left
Explore available promotions

Description

Description

Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

Solubility
Soluble in ethanol and water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
Specifications

Specifications

Manganese(II) nitrate tetrahydrate
20694-39-7
H8MnN2O10
MFCD00149789
14,5733
Soluble in ethanol and water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]
251.006
251.01 (178.95 Anhydrous)
Odorless
∼26°C
1000 g
Mn(NO3)2·4H2O
UN2724
manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
ALIMWUQMDCBYFM-UHFFFAOYSA-N
manganese(2+);dinitrate;tetrahydrate
182616
98%
Hygroscopic

Chemical Identifiers

20694-39-7
251.006
ALIMWUQMDCBYFM-UHFFFAOYSA-N
182616
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]
H8MnN2O10
MFCD00149789
manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
manganese(2+);dinitrate;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318-H373-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H272-H302-H314-H373

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: MANGANESE NITRATE

EINECSNumber : 233-828-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only