missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) fluoride, 99%
Description
Manganese(II) fluoride is used in the manufacture of special kinds of glass and lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 856°C |
| Color | Red |
| Physical Form | Crystalline Powder |
| Quantity | 10 g |
| Assay Percent Range | 99% |
| Solubility Information | Insoluble in water,alcohol. Soluble in dilute HF,conc. HCl or HNO3 |
| Formula Weight | 92.93 |
| Sensitivity | Hygroscopic |
| Density | 3.98 |
| Chemical Name or Material | Manganese(II) fluoride |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 231-960-0 |
| RTECSNumber | OP0875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only