missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Tetrahydrate 98.0+%, TCI America™

Catalog No. M2095500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2095500G 500 g
M209525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2095500G Supplier TCI America Supplier No. M2095500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 650°C
Color Red-White
Physical Form Crystalline Powder
Quantity 500 g
UN Number 3288
Formula Weight 125.84
Percent Purity ≥98.0% (T)
Grade Reagent
Chemical Name or Material Manganese(II) Chloride Tetrahydrate
EINECSNumber (1)-0235
RTECSNumber OO9625000
TSCA Yes