missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Chloride Tetrahydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 650°C |
| Color | Red-White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| UN Number | 3288 |
| Formula Weight | 125.84 |
| Percent Purity | ≥98.0% (T) |
| Grade | Reagent |
| Chemical Name or Material | Manganese(II) Chloride Tetrahydrate |
Safety and Handling
| EINECSNumber | (1)-0235 |
| RTECSNumber | OO9625000 |
| TSCA | Yes |