missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1080418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1080418 50 g
AA1080409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1080418 Supplier Thermo Scientific Chemicals Supplier No. 01080418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

Manganese chloride is used as a source of manganese ion in biological research, such as in studies of manganese uptake in rat liver. It has been substituted for magnesium chloride to investigate alterations in activity of the Herpes simplex virus type 1 DNA polymerase. It is also used in industrial applications such as dyeing and disinfecting, the purification of natural gas, and as a drying agent for linseed oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Physical Form Crystalline Powder
Quantity 50 g
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 99.999% (Metals basis)
Density 2.01
Solubility Information Soluble in water, pyridine and ethanol. Insoluble in ether.
Formula Weight 197.90 (125.84 Anhydrous)
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less
Hazard Category H302-H318-H373
Hazard Statement Wear suitable protective clothing.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-869-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only