missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(Ii) Chloride Tetrahydrate, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Packaging | Plastic Bottle |
| Linear Formula | MnCl2 · 4H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 197.91g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Manganese(Ii) Chloride Tetrahydrate |
Safety and Handling
| Hazard Statement | P273 |
| Precautionary Statement | H302-H412 |
| EINECSNumber | 231-869-6 |
| RTECSNumber | OO9650000 |