missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

Supplier:  Thermo Scientific Chemicals 03652636

 View more versions of this product

Catalog No. AA3652636


Only null left
Explore available promotions

Description

Description

It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(II) chloride tetrahydrate
13446-34-9
Crystalline
98-101%
MFCD00149792
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+) tetrahydrate dichloride
197.90 (125.84 Anhydrous)
Odorless
Pink crystalline
58°C
Pink
500 g
Cl2H8MnO4
Soluble in water,pyridine and ethanol. Insoluble in ether.
O.O.O.O.[Cl-].[Cl-].[Mn++]
197.90
ACS
Hygroscopic
2.01

Chemical Identifiers

13446-34-9
197.90
CNFDGXZLMLFIJV-UHFFFAOYSA-L
O.O.O.O.[Cl-].[Cl-].[Mn++]
Cl2H8MnO4
MFCD00149792
manganese(2+) tetrahydrate dichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing.

P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-869-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only