missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Description
It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 58°C |
| Color | Pink |
| Physical Form | Crystalline |
| Quantity | 2 kg |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Pink crystalline |
| Assay Percent Range | 98-101% |
| Density | 2.010 g/cm3 at 20°C |
| Solubility Information | Soluble in water,pyridine and ethanol. Insoluble in ether. |
| Show More |
Safety and Handling
| Hazard Category | H302-H318-H373 |
| Hazard Statement | Wear suitable protective clothing. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 231-869-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only