missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride tetrahydrate, 99% (metals basis)
Description
Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 58°C |
| Color | Pink |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Assay Percent Range | 99% (Metal basis) |
| Solubility Information | Soluble in pyridine,ethanol. Insoluble in ether. |
| Formula Weight | 197.90 (125.84 Anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Pink Crystalline/powder |
| Show More |
Safety and Handling
| Hazard Category | H302-H318-H373 |
| Precautionary Statement | P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 231-869-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only