missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, 99% (metals basis)

Catalog No. AA1156322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156322 100 g
AA1156336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1156322 Supplier Thermo Scientific Chemicals Supplier No. 01156322

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% (Metal basis)
Solubility Information Soluble in pyridine,ethanol. Insoluble in ether.
Formula Weight 197.90 (125.84 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Pink Crystalline/powder
Density 2.010 g/cm3 at 20°C
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less
Hazard Category H302-H318-H373
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-869-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only