missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, 1M aq. soln.

Catalog No. p-7058236
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63150AD 50 mL
AAJ63150AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63150AD Supplier Thermo Scientific Chemicals Supplier No. J63150AD
Only null left

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
IUPAC Name manganese(2+) dichloride
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula MnCl2
Formula Weight 125.84
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Manganese(II) chloride
Hazard Category H318-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P260-P280-P305+P351+P338-P310-P314-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only