Learn More
Manganese(II) carbonate, Puratronic™, 99.985% (metals basis)
CAS: 598-62-9 | CMnO3 | 114.946 g/mol
Supplier: Thermo Scientific Chemicals 04075614
Description
Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Manganese(II) carbonate | |
| Brown to Violet | |
| Powder | |
| 99.99% (metals basis) | |
| MFCD00011116 | |
| Soluble in water(0.065g/L),dilute inorganic acids. Insoluble alcohol. | |
| C(=O)([O-])[O-].[Mn+2] | |
| 114.946 | |
| 114.95 | |
| Light brown to violet powder |
| 350°C (decomposition) | |
| 598-62-9 | |
| 25 g | |
| CMnO3 | |
| manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660 | |
| XMWCXZJXESXBBY-UHFFFAOYSA-L | |
| manganese(2+);carbonate | |
| 11726 | |
| Odorless | |
| 3.125 |
Chemical Identifiers
| 598-62-9 | |
| 114.946 | |
| XMWCXZJXESXBBY-UHFFFAOYSA-L | |
| 11726 | |
| C(=O)([O-])[O-].[Mn+2] |
| CMnO3 | |
| MFCD00011116 | |
| manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660 | |
| manganese(2+);carbonate |
Safety and Handling
EINECSNumber : 209-942-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only