missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis)

Catalog No. AA4075622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4075622 100 g
AA4075606 5 g
AA4075614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4075622 Supplier Thermo Scientific Chemicals Supplier No. 04075622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-62-9 | CMnO3 | 114.946 g/mol

Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-62-9
Molecular Formula CMnO3
Molecular Weight (g/mol) 114.946
MDL Number MFCD00011116
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
Synonym manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
PubChem CID 11726
IUPAC Name manganese(2+);carbonate
SMILES C(=O)([O-])[O-].[Mn+2]

Specifications

Melting Point 350°C (decomposition)
Color Brown to Violet
Physical Form Powder
Quantity 100 g
Odor Odorless
Appearance Light brown to violet powder
Assay Percent Range 99.99% (metals basis)
Density 3.125
Solubility Information Soluble in water(0.065g/L),dilute inorganic acids. Insoluble alcohol.
Formula Weight 114.95
Chemical Name or Material Manganese(II) carbonate
Show More Show Less
EINECSNumber 209-942-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only